C12H316CF41A Chemcia Scientific, LLC

5-Bromo-1-oxo-1,3-dihydro-isoindole-2-carboxylic acid tert-butyl ester

  • MDL number MFCD17392825
  • Cas Number : 864866-80-8
  • Molecular Weight : 312.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 5-bromo-1-oxo-1,3-dihydro-2H-isoindole-2-carboxylate
Inchi Code 1S/C13H14BrNO3/c1-13(2,3)18-12(17)15-7-8-6-9(14)4-5-10(8)11(15)16/h4-6H,7H2,1-3H3
Inchi Key AYNRRKAUKGRPMZ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9071

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9071
Availability details for
Quantity
  • Availability
  • Add to Cart