C12H316CF41C Chemcia Scientific, LLC

5-Chloro-isophthalic acid monomethyl ester

  • MDL number MFCD08458982
  • Cas Number : 153203-57-7
  • Molecular Weight : 214.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-5-(methoxycarbonyl)benzoic acid
Inchi Code 1S/C9H7ClO4/c1-14-9(13)6-2-5(8(11)12)3-7(10)4-6/h2-4H,1H3,(H,11,12)
Inchi Key HLARAVKGVUMKCY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9084

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9084
Availability details for
Quantity
  • Availability
  • Add to Cart