C12H316CF41D Chemcia Scientific, LLC

1-(2-Fluoro-4-methoxy-5-nitro-phenyl)-ethanone

  • MDL number MFCD30723716
  • Cas Number : 1414877-12-5
  • Molecular Weight : 213.16681

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9088

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9088
Availability details for
Quantity
  • Availability
  • Add to Cart