C12H316CF420 Chemcia Scientific, LLC

6-Hydroxy-3-methyl-indazole-1-carboxylic acid tert-butyl ester

  • MDL number MFCD24467331
  • Cas Number : 1055974-00-9
  • Molecular Weight : 248.28407

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9093

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9093
Availability details for
Quantity
  • Availability
  • Add to Cart