C12H316CF421 Chemcia Scientific, LLC

6-Hydroxy-3-methyl-benzo[b]thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD29373040
  • Cas Number : 82788-19-0
  • Molecular Weight : 222.26455

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9094

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9094
Availability details for
Quantity
  • Availability
  • Add to Cart