C12H316CF422 Chemcia Scientific, LLC

6-Amino-2-trifluoromethyl-nicotinonitrile

  • MDL number MFCD11848372
  • Cas Number : 1233243-98-5
  • Molecular Weight : 187.12523

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9096

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9096
Availability details for
Quantity
  • Availability
  • Add to Cart