C12H316CF424 Chemcia Scientific, LLC

4-Chloro-[1,5]naphthyridine-3-carboxylic acid ethyl ester

  • MDL number MFCD26526928
  • Cas Number : 83067-95-2
  • Molecular Weight : 236.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-chloro-1,5-naphthyridine-3-carboxylate
Inchi Code 1S/C11H9ClN2O2/c1-2-16-11(15)7-6-14-8-4-3-5-13-10(8)9(7)12/h3-6H,2H2,1H3
Inchi Key ZAUJIZUBUCDAEU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9114

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9114
Availability details for
Quantity
  • Availability
  • Add to Cart