C12H316CF425 Chemcia Scientific, LLC

4-Chloro-6-methoxy-[1,5]naphthyridine-3-carboxylic acid ethyl ester

  • MDL number MFCD29053301
  • Cas Number : 162220-96-4
  • Molecular Weight : 266.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-chloro-6-methoxy-1,5-naphthyridine-3-carboxylate
Inchi Code 1S/C12H11ClN2O3/c1-3-18-12(16)7-6-14-8-4-5-9(17-2)15-11(8)10(7)13/h4-6H,3H2,1-2H3
Inchi Key GOHQELXFJUGXNN-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9123

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9123
Availability details for
Quantity
  • Availability
  • Add to Cart