C12H316CF428 Chemcia Scientific, LLC

(3,5-Difluoro-pyridin-4-yl)-methanol

  • MDL number MFCD16606966
  • Cas Number : 924649-16-1
  • Molecular Weight : 145.10965

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9155

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9155
Availability details for
Quantity
  • Availability
  • Add to Cart