C12H316CF42B Chemcia Scientific, LLC

7-Chloro-2H-pyrazolo[3,4-c]pyridine

  • MDL number MFCD27988501
  • Cas Number : 76006-11-6
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-chloro-2H-pyrazolo[3,4-c]pyridine
Inchi Code 1S/C6H4ClN3/c7-6-5-4(1-2-8-6)3-9-10-5/h1-3H,(H,9,10)
Inchi Key LGYXPMDWRGMZCV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9161

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9161
Availability details for
Quantity
  • Availability
  • Add to Cart