C12H316CF42E Chemcia Scientific, LLC

7-Methoxy-2H-pyrazolo[3,4-c]pyridine

  • MDL number MFCD30723740
  • Molecular Weight : 149.15334

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9164

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9164
Availability details for
Quantity
  • Availability
  • Add to Cart