C12H316CF42F Chemcia Scientific, LLC

1,6-Dihydro-pyrazolo[3,4-c]pyridin-7-one

  • MDL number MFCD11846326
  • Cas Number : 76006-09-2
  • Molecular Weight : 135.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,6-dihydro-7H-pyrazolo[3,4-c]pyridin-7-one
Inchi Code 1S/C6H5N3O/c10-6-5-4(1-2-7-6)3-8-9-5/h1-3H,(H,7,10)(H,8,9)
Inchi Key RPXFCERITFUFCN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9171

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9171
Availability details for
Quantity
  • Availability
  • Add to Cart