C12H316CF432 Chemcia Scientific, LLC

3-Bromo-1,6-dihydro-pyrazolo[3,4-c]pyridin-7-one

  • MDL number MFCD09265726
  • Cas Number : 956077-63-7
  • Molecular Weight : 214.02228

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9177

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9177
Availability details for
Quantity
  • Availability
  • Add to Cart