C12H316CF437 Chemcia Scientific, LLC

6-Bromo-8-methoxy-imidazo[1,2-a]pyridine

  • MDL number MFCD23708758
  • Cas Number : 1427424-35-8
  • Molecular Weight : 227.06179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9211

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9211
Availability details for
Quantity
  • Availability
  • Add to Cart