C12H316CF43A Chemcia Scientific, LLC

7-Chloro-3-nitro-1H-pyrazolo[3,4-c]pyridine

  • MDL number MFCD09265715
  • Cas Number : 957760-19-9
  • Molecular Weight : 198.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-chloro-3-nitro-1H-pyrazolo[3,4-c]pyridine
Inchi Code 1S/C6H3ClN4O2/c7-5-4-3(1-2-8-5)6(10-9-4)11(12)13/h1-2H,(H,9,10)
Inchi Key PSJJYTFYDCJOFS-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9248

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9248
Availability details for
Quantity
  • Availability
  • Add to Cart