C12H316CF43B Chemcia Scientific, LLC

7-Chloro-1H-pyrazolo[3,4-c]pyridin-3-ylamine

  • MDL number MFCD30834093
  • Molecular Weight : 168.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-chloro-1H-pyrazolo[3,4-c]pyridin-3-amine
Inchi Code 1S/C6H5ClN4/c7-5-4-3(1-2-9-5)6(8)11-10-4/h1-2H,(H3,8,10,11)
Inchi Key LPAZQMHJRQTIIQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9249

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9249
Availability details for
Quantity
  • Availability
  • Add to Cart