C12H316CF43D Chemcia Scientific, LLC

3-Amino-4-methyl-pyridine-2-carboxylic acid methyl ester

  • MDL number MFCD22551647
  • Cas Number : 1263059-97-7
  • Molecular Weight : 166.1811

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9260

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9260
Availability details for
Quantity
  • Availability
  • Add to Cart