C12H316CF43E Chemcia Scientific, LLC

3-Amino-4-methyl-pyridine-2-carboxylic acid

  • MDL number MFCD13174786
  • Cas Number : 53636-30-9
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-4-methyl-2-pyridinecarboxylic acid
Inchi Code 1S/C7H8N2O2/c1-4-2-3-9-6(5(4)8)7(10)11/h2-3H,8H2,1H3,(H,10,11)
Inchi Key FDZGMCXGIXQJPA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9261

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9261
Availability details for
Quantity
  • Availability
  • Add to Cart