C12H316CF441 Chemcia Scientific, LLC

6-Bromo-8-methoxy-[1,2,4]triazolo[1,5-a]pyrazine

  • MDL number MFCD30834113
  • Cas Number : 2169539-35-7
  • Molecular Weight : 229.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-8-methoxy-[1,2,4]triazolo[1,5-a]pyrazine
Inchi Code 1S/C6H5BrN4O/c1-12-6-5-8-3-9-11(5)2-4(7)10-6/h2-3H,1H3
Inchi Key WFWZVVBGEDQSKX-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9293

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9293
Availability details for
Quantity
  • Availability
  • Add to Cart