C12H316CF442 Chemcia Scientific, LLC

6-Chloro-8-methoxy-[1,2,4]triazolo[1,5-a]pyrazine

  • MDL number MFCD30834124
  • Molecular Weight : 184.58595

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9309

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9309
Availability details for
Quantity
  • Availability
  • Add to Cart