C12H316CF445 Chemcia Scientific, LLC

5-Chloro-6-methoxy-2,3-dihydro-isoindol-1-one

  • MDL number MFCD21362709
  • Cas Number : 1138220-79-7
  • Molecular Weight : 197.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-6-methoxy-1-isoindolinone
Inchi Code 1S/C9H8ClNO2/c1-13-8-3-6-5(2-7(8)10)4-11-9(6)12/h2-3H,4H2,1H3,(H,11,12)
Inchi Key MMBRLXRMVPEDCW-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9334

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9334
Availability details for
Quantity
  • Availability
  • Add to Cart