C12H316CF448 Chemcia Scientific, LLC

7-Chloro-2H-pyrazolo[3,4-c]pyridin-3-ylamine

  • MDL number MFCD30834140
  • Molecular Weight : 168.58655

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9353

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9353
Availability details for
Quantity
  • Availability
  • Add to Cart