C12H316CF44F Chemcia Scientific, LLC

3-Methoxy-4-methyl-2-nitro-benzoic acid

  • MDL number MFCD09834816
  • Cas Number : 57281-77-3
  • Molecular Weight : 211.17578

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9356

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9356
Availability details for
Quantity
  • Availability
  • Add to Cart