C12H316CF450 Chemcia Scientific, LLC

2-Bromo-6-methoxy-4-methyl-pyridin-3-ylamine

  • MDL number MFCD09839257
  • Cas Number : 135795-51-6
  • Molecular Weight : 217.06658

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9360

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9360
Availability details for
Quantity
  • Availability
  • Add to Cart