C12H316CF453 Chemcia Scientific, LLC

1-(5-Bromo-2-fluoro-4-methyl-phenyl)-ethanone

  • MDL number MFCD28397870
  • Cas Number : 1785558-45-3
  • Molecular Weight : 231.06591

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9382

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9382
Availability details for
Quantity
  • Availability
  • Add to Cart