C12H316CF455 Chemcia Scientific, LLC

7-Fluoro-1H-[1,5]naphthyridin-4-one

  • MDL number MFCD28098173
  • Cas Number : 1151802-05-9
  • Molecular Weight : 164.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-fluoro-1,5-naphthyridin-4-ol
Inchi Code 1S/C8H5FN2O/c9-5-3-6-8(11-4-5)7(12)1-2-10-6/h1-4H,(H,10,12)
Inchi Key XPJNOFYNSDLDOF-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9424

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9424
Availability details for
Quantity
  • Availability
  • Add to Cart