C12H316CF459 Chemcia Scientific, LLC

6-Methoxy-1H-cinnolin-4-one

  • MDL number MFCD24691764
  • Molecular Weight : 176.17631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9499

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9499
Availability details for
Quantity
  • Availability
  • Add to Cart