C12H316CF45C Chemcia Scientific, LLC

5-Fluoro-2H-indazol-3-ylamine

  • MDL number MFCD20489387
  • Molecular Weight : 151.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-2H-indazol-3-amine
Inchi Code 1S/C7H6FN3/c8-4-1-2-6-5(3-4)7(9)11-10-6/h1-3H,(H3,9,10,11)
Inchi Key UWLBVJRYQQUECL-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9507

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9507
Availability details for
Quantity
  • Availability
  • Add to Cart