C12H316CF460 Chemcia Scientific, LLC

2-Amino-4-bromo-5-methyl-benzoic acid

  • Cas Number : 1643156-29-9
  • Molecular Weight : 230.06246

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9536

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9536
Availability details for
Quantity
  • Availability
  • Add to Cart