C12H316CF465 Chemcia Scientific, LLC

1-Chloro-5-iodo-4-methoxy-2-methyl-benzene

  • MDL number MFCD27929828
  • Cas Number : 850864-46-9
  • Molecular Weight : 282.50976

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 93
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9576

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9576
Availability details for
Quantity
  • Availability
  • Add to Cart