C12H316CF470 Chemcia Scientific, LLC

6-Methyl-1H-[1,5]naphthyridin-4-one

  • MDL number MFCD18828523
  • Cas Number : 724787-72-8
  • Molecular Weight : 160.17691

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9606

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9606
Availability details for
Quantity
  • Availability
  • Add to Cart