C12H316CF476 Chemcia Scientific, LLC

6-Bromo-imidazo[1,2-a]pyrazine-8-carboxylic acid

  • MDL number MFCD13189932
  • Cas Number : 1196154-13-8
  • Molecular Weight : 242.03283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9703

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9703
Availability details for
Quantity
  • Availability
  • Add to Cart