C12H316CF478 Chemcia Scientific, LLC

8-Bromo-imidazo[1,2-a]pyrazine-6-carboxylic acid methyl ester

  • MDL number MFCD30803878
  • Cas Number : 1660116-54-0
  • Molecular Weight : 256.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 8-bromoimidazo[1,2-a]pyrazine-6-carboxylate
Inchi Code 1S/C8H6BrN3O2/c1-14-8(13)5-4-12-3-2-10-7(12)6(9)11-5/h2-4H,1H3
Inchi Key NZEVGXNKWJPIHR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9711

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9711
Availability details for
Quantity
  • Availability
  • Add to Cart