C12H316CF479 Chemcia Scientific, LLC

5-Amino-6-chloro-pyrazine-2-carboxylic acid methyl ester

  • MDL number MFCD16657583
  • Cas Number : 1378678-66-0
  • Molecular Weight : 187.58662

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9712

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9712
Availability details for
Quantity
  • Availability
  • Add to Cart