C12H316CF48B Chemcia Scientific, LLC

5-Amino-1H-indazole-3-carbonitrile

  • MDL number MFCD07371583
  • Cas Number : 89939-59-3
  • Molecular Weight : 158.16382

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9805

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9805
Availability details for
Quantity
  • Availability
  • Add to Cart