C12H316CF48E Chemcia Scientific, LLC

5-Nitro-2H-indazole-3-carbonitrile

  • MDL number MFCD28158908
  • Molecular Weight : 188.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-nitro-2H-indazole-3-carbonitrile
Inchi Code 1S/C8H4N4O2/c9-4-8-6-3-5(12(13)14)1-2-7(6)10-11-8/h1-3H,(H,10,11)
Inchi Key WXTZAAHDBOKXDI-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin CN
Product Link http://www.chemcia.com/product_fl02.asp?id=9808

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9808
Availability details for
Quantity
  • Availability
  • Add to Cart