C12H316CF495 Chemcia Scientific, LLC

5-Amino-2-methyl-4-nitro-benzoic acid methyl ester

  • MDL number MFCD18642362
  • Cas Number : 116448-91-0
  • Molecular Weight : 210.19105

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9850

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9850
Availability details for
Quantity
  • Availability
  • Add to Cart