C12H316CF498 Chemcia Scientific, LLC

4-Amino-2-chloro-5-nitro-benzoic acid

  • Cas Number : 82378-89-0
  • Molecular Weight : 216.5819

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9926

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9926
Availability details for
Quantity
  • Availability
  • Add to Cart