C12H316CF499 Chemcia Scientific, LLC

4-Amino-2-bromo-5-nitro-benzoic acid

  • MDL number MFCD17168934
  • Cas Number : 860182-17-8
  • Molecular Weight : 261.0329

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9928

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9928
Availability details for
Quantity
  • Availability
  • Add to Cart