C12H316CF49C Chemcia Scientific, LLC

6-Bromo-3-chloro-1H-pyrrolo[3,2-b]pyridine

  • Cas Number : 1190319-44-8
  • Molecular Weight : 231.48033

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9965

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9965
Availability details for
Quantity
  • Availability
  • Add to Cart