C12H316CF49F Chemcia Scientific, LLC

2-Amino-4-bromo-3-chloro-phenol

  • MDL number MFCD30834309
  • Cas Number : 2091488-61-6
  • Molecular Weight : 222.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-4-bromo-3-chlorophenol
Inchi Code 1S/C6H5BrClNO/c7-3-1-2-4(10)6(9)5(3)8/h1-2,10H,9H2
Inchi Key SDFYQMCRZVAVAK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9979

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9979
Availability details for
Quantity
  • Availability
  • Add to Cart