C12H316CF4C1 Chemcia Scientific, LLC

3-Amino-pyridin-2-ol hydrochloride

  • MDL number MFCD15209580
  • Cas Number : 33631-21-9
  • Molecular Weight : 146.57734

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Black Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10039

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10039
Availability details for
Quantity
  • Availability
  • Add to Cart