C12H316CF4C3 Chemcia Scientific, LLC

5-Chloro-2H-indazole-7-carboxylic acid methyl ester

  • MDL number MFCD28159290
  • Molecular Weight : 210.62134

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10074

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10074
Availability details for
Quantity
  • Availability
  • Add to Cart