C12H316CF4C6 Chemcia Scientific, LLC

5-Chloro-2H-indazole-7-carboxylic acid

  • MDL number MFCD28159289
  • Molecular Weight : 196.59425

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10077

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10077
Availability details for
Quantity
  • Availability
  • Add to Cart