C12H316CF4CD Chemcia Scientific, LLC

5-Fluoro-1H-indazole-7-carboxylic acid

  • MDL number MFCD27987844
  • Cas Number : 1038916-13-0
  • Molecular Weight : 180.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-1H-indazole-7-carboxylic acid
Inchi Code 1S/C8H5FN2O2/c9-5-1-4-3-10-11-7(4)6(2-5)8(12)13/h1-3H,(H,10,11)(H,12,13)
Inchi Key MRPARTBZLWCDIU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10093

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10093
Availability details for
Quantity
  • Availability
  • Add to Cart