C12H316CF4CF Chemcia Scientific, LLC

4-Bromo-3-chloro-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD11046241
  • Cas Number : 943323-92-0
  • Molecular Weight : 231.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-3-chloro-1H-pyrrolo[2,3-b]pyridine
Inchi Code 1S/C7H4BrClN2/c8-4-1-2-10-7-6(4)5(9)3-11-7/h1-3H,(H,10,11)
Inchi Key NTAZESWKVUMQKW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10101

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10101
Availability details for
Quantity
  • Availability
  • Add to Cart