C12H316CF4D1 Chemcia Scientific, LLC

4-Bromo-2-chloro-5-fluoro-benzonitrile

  • MDL number MFCD11036199
  • Cas Number : 1126779-33-6
  • Molecular Weight : 234.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2-chloro-5-fluorobenzonitrile
Inchi Code 1S/C7H2BrClFN/c8-5-2-6(9)4(3-11)1-7(5)10/h1-2H
Inchi Key QRVZOLCBNJYMJK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10120

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10120
Availability details for
Quantity
  • Availability
  • Add to Cart