C12H316CF4D2 Chemcia Scientific, LLC

5-Amino-2-iodo-4-methyl-benzoic acid methyl ester

  • Cas Number : 2092115-73-4
  • Molecular Weight : 291.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-amino-2-iodo-4-methylbenzoate
Inchi Code 1S/C9H10INO2/c1-5-3-7(10)6(4-8(5)11)9(12)13-2/h3-4H,11H2,1-2H3
Inchi Key VIQSHNCWSNFRSX-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10137

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10137
Availability details for
Quantity
  • Availability
  • Add to Cart