C12H316CF4DB Chemcia Scientific, LLC

2-Bromo-4-methyl-5-nitro-benzoic acid methyl ester

  • MDL number MFCD28745250
  • Cas Number : 1202858-67-0
  • Molecular Weight : 274.07241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10162

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10162
Availability details for
Quantity
  • Availability
  • Add to Cart