C12H316CF4DC Chemcia Scientific, LLC

5-Amino-2-fluoro-4-methyl-benzonitrile

  • MDL number MFCD28131807
  • Cas Number : 1426136-04-0
  • Molecular Weight : 150.15679

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10163

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10163
Availability details for
Quantity
  • Availability
  • Add to Cart